Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human/Mouse STAT4, Polyclonal, R&D Systems™

Rabbit Polyclonal Antibody

Manufacturer:  R&D Systems PAST4

Catalog No. PAST4


Add to cart
This item is not returnable. View return policy

Description

Description

Host Species: Rabbit
Species Reactivity:Human, Mouse
Immunogen:aa 730 - 749 of murine STAT4

Applications:

Immunoprecipitation, Western Blotting

Specifications

Specifications

STAT4
Unconjugated
Lyophilized from 0.2 mL antiserum containing 0.02% sodium azide. with 0.02% Sodium Azide
signal transducer and activator of transcription 4, SLEB11
Rabbit
Serum
RUO
Primary
6775
Human, Mouse
Immunoprecipitation, Western Blot
Western Blot 1:2000 dilution, Immunoprecipitation 3 ug/500 ug cell lysate
Serum
STAT4
aa 730 - 749 of murine STAT4
200 uL
Store the unopened product at -20 to -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Polyclonal
Detects human and murine STAT4. Does not cross-react with any of the other STAT proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit