Learn More
Tmp21/p23 Sheep anti-Human, Mouse, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF7630
Catalog No. AF7630
Add to cart
This item is not returnable. View return policy
Description
Description
Tmp21/p23 Polyclonal antibody AF7630 Specifically detects Tmp21/p23 in Human, Mouse samples. It is validated for Western Blotting
Specifications
Specifications
Tmp21/p23
LYOPH
Western Blot 0.5 ug/mL, Immunohistochemistry 1-15 ug/mL, Knockout Validated
P49755
21 kDa transmembrane-trafficking protein, p23, P24(DELTA), p24delta, S31I125, S31III125, TMED10, TMP21Tmp-21-I, transmembrane emp24 domain-containing protein 10, transmembrane emp24-like trafficking protein 10 (yeast), Transmembrane protein Tmp21,21 kDa transmembrane trafficking protein
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse in direct ELISAs and Western blots.
Human, Mouse
Immunoassay, Immunohistochemistry, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TMED10
E. coli-derived recombinant human Tmp21/p23 Ile32-Arg185 Accession # P49755
Immunogen affinity purified
RUO
Primary
10972
Sterile PBS to a final concentration of 0.2 mg/mL.