Learn More
PAK1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF7495
Catalog No. AF7495
Add to cart
This item is not returnable. View return policy
Description
Description
PAK1 Polyclonal antibody AF7495 Specifically detects PAK1 in Human samples. It is validated for Immunocytochemistry
Specifications
Specifications
PAK1
LYOPH
Immunocytochemistry 5-15 ug/mL
Q13153
alpha-PAK, EC 2.7.11, EC 2.7.11.1, MGC130000, MGC130001, p21 protein (Cdc42/Rac)-activated kinase 1, p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast), p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related), p21-activated kinase 1, p65-PAK, PAK-1, PAKalpha, serine/threonine-protein kinase PAK 1, STE20 homolog, yeast
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human PAK1 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) PAK1B, rhPAK2, rhPAK4, rhPAK6, and rhPAK7 is observed.
Human
Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PAK1
E. coli-derived recombinant human PAK1 Leu128-Gln242 Accession # Q13153
Immunogen affinity purified
RUO
Primary
5058
Sterile PBS to a final concentration of 0.2 mg/mL.