Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Prox1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 61 publication
Supplier: R&D Systems AF2727
This item is not returnable.
View return policy
Description
Prox1 Polyclonal specifically detects Prox1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
Prox1 | |
Polyclonal | |
Unconjugated | |
Q92786 | |
PROX1 | |
E. coli-derived recombinant human Prox1 Pro2-Gln259 Accession # Q92786 | |
100 μg | |
Primary | |
Detects human Prox1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Homeobox prospero-like protein PROX1, prospero homeobox 1, prospero homeobox protein 1, prospero-related homeobox 1, PROX-1 | |
Goat | |
Affinity Purified | |
RUO | |
5629 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title