Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PSMB10/MECL1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF7684
This item is not returnable.
View return policy
Description
PSMB10/MECL1 Polyclonal specifically detects PSMB10/MECL1 in Human samples. It is validated for Western Blot.Specifications
PSMB10/MECL1 | |
Polyclonal | |
Unconjugated | |
P40306 | |
PSMB10 | |
E. coli-derived recombinant human PSMB10/MECL1 Thr40-Glu273 Accession # P40306 | |
100 μg | |
Primary | |
Detects human PSMB10/MECL1 in direct ELISAs and Western blots. In direct ELISAs, less than 6% cross-reactivity with recombinant human PSMB7 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
beta2i, FLJ00366, LMP10, LMP10Proteasome subunit beta-2i, Low molecular mass protein 10, Macropain subunit MECl-1, MECl-1, MECL1, MECL1EC 3.4.25.1, MGC1665, Multicatalytic endopeptidase complex subunit MECl-1, proteasome (prosome, macropain) subunit, beta type, 10, proteasome catalytic subunit 2i, Proteasome MECl-1, proteasome subunit beta 7i, proteasome subunit beta type-10, Proteasome subunit beta-2i, proteasome subunit MECL1 | |
Goat | |
Affinity Purified | |
RUO | |
5699 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction