Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PTP sigma/PTPRS Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Supplier: R&D Systems AF3430
This item is not returnable.
View return policy
Description
PTP sigma/PTPRS Polyclonal specifically detects PTP sigma/PTPRS in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
PTP sigma/PTPRS | |
Polyclonal | |
Unconjugated | |
NP_570924 | |
PTPRS | |
Mouse myeloma cell line NS0-derived recombinant human PTP sigma/PTPRS Glu30-Gly1260 Accession # NP_570924 | |
100 μg | |
Primary | |
Detects the endogenous cleaved extracellular domain of human PTP sigma/PTPRS in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human LAR, PTPR beta , PTPR delta , PTPRμ, or PTPR kappa is observed. | |
Human | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
EC 3.1.3.48, protein tyrosine phosphatase PTPsigma, protein tyrosine phosphatase, receptor type, S, protein tyrosine phosphatase, receptor type, sigma, PTPR sigma, PTPRS, PTPSIGMA, receptor-type tyrosine-protein phosphatase S, Receptor-type tyrosine-protein phosphatase sigma, R-PTP-S, R-PTP-sigma | |
Goat | |
Affinity Purified | |
RUO | |
5802 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction