Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GDNF Goat anti-Human, Rat, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Supplier: R&D Systems AF212NA
This item is not returnable.
View return policy
Description
GDNF Polyclonal specifically detects GDNF in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
GDNF | |
Polyclonal | |
Unconjugated | |
P39905 | |
GDNF | |
E. coli-derived and Mouse myeloma cell line NS0-derived recombinant human GDNF Arg109-Ile211 Accession # P39905 | |
100 μg | |
Primary | |
Detects human and rat GDNF in direct ELISAs and Western blots. | |
Human, Rat | |
IgG |
Western Blot, Immunohistochemistry, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization 0.25-1 ug/mL | |
Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3 | |
Goat | |
Affinity Purified | |
RUO | |
2668 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title