Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GFAP Sheep anti-Human, Rat, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody has been used in 5 publications
Supplier: R&D Systems AF2594
This item is not returnable.
View return policy
Description
GFAP Polyclonal specifically detects GFAP in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Specifications
GFAP | |
Polyclonal | |
Unconjugated | |
P14136 | |
GFAP | |
E. coli-derived recombinant human GFAP Leu292-Met432 Accession # P14136 | |
Affinity Purified | |
RUO | |
2670 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.2 ug/mL, Simple Western 0.1-2 ug/mL, Immunocytochemistry 5-15 ug/mL | |
FLJ45472, GFAP astrocytes, glial fibrillary acidic protein | |
Sheep | |
50 | |
100 μg | |
Primary | |
Detects human and rat GFAP in Western blots. | |
Human, Rat | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title