Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Human Serum Albumin, Clone: 15C7, Abcam™
Supplier: Abcam AB10241250UG
Description
Host Species: Mouse
Clone: 15C7
Isotype: IgG2b
Cross Reactivity: Human. Does not react with Chicken, Cow
Immunogen: Human serum albumin.
Applications:
ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blotting
Specifications
Human Serum Albumin | |
15C7 | |
Unconjugated | |
Human serum albumin. | |
Cancer and Tumor Biology, Cardiovascular, Stem Cell Biology | |
Human | |
IgG2b |
Monoclonal | |
2mg/mL | |
Mouse | |
250 μg | |
Primary | |
Purified |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction