Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SH2D1A Sheep anti-Human, Polyclonal, R&D Systems
Sheep Polyclonal Antibody
Manufacturer: R&D Systems AF7440
This item is not returnable.
View return policy
Description
SH2D1A Polyclonal antibody AF7440 Specifically detects SH2D1A in Human samples. It is validated for Western BlottingSpecifications
SH2D1A | |
LYOPH | |
Western Blot 1 ug/mL | |
O60880 | |
DSHP, DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, LYP, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLP, XLPD, XLPDFLJ18687, XLPSH2 domain protein 1A | |
Sheep | |
IgG | |
100 ug | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
Polyclonal | |
Detects human SH2D1A in direct ELISAs and Western blots. | |
Human |
Western Blot | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Affinity Purified | |
SH2D1A | |
E. coli-derived recombinant human SH2D1A Met1-Pro128 Accession # O60880 | |
Immunogen affinity purified | |
RUO | |
Primary | |
4068 | |
Sterile PBS to a final concentration of 0.2 mg/mL. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title