Learn More
SOX21 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Manufacturer:  R&D Systems AF3538
Catalog No. AF3538
Add to cart
This item is not returnable. View return policy
Description
Description
SOX21 Polyclonal antibody AF3538 Specifically detects SOX21 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
SOX21
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
Q9Y651
SOX25, SOX25SOX-A, SOXA, SRY (sex determining region Y)-box 21, SRY-box 21, transcription factor SOX-21
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human SOX21 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) SOX1, rhSOX2 and rhSOX15 is observed.
Human
Immunocytochemistry, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SOX21
E. coli-derived recombinant human SOX21 Ser2-Leu276 (Gly102Ser) Accession # Q9Y651
Immunogen affinity purified
RUO
Primary
11166
Reconstitute at 0.2 mg/mL in sterile PBS.