Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human STAT2, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Manufacturer:  R&D Systems PAFST2

Catalog No. PAFST2


Add to cart
This item is not returnable. View return policy

Description

Description

Host Species: Goat
Species Reactivity:Human
Immunogen:aa 679 - 851 of human STAT2

Applications:

Immunoprecipitation, Western Blotting

Specifications

Specifications

STAT2
Unconjugated
Lyophilized from a 0.2 μm sterile-filtered solution in phosphate-buffered saline (PBS) with 5% trehalose. with No Preservative
Affinity Purified
STAT2
E. coli-derived recombinant human STAT2 Gln679-Phe851 Accession # P52630
Antigen Affinity-purified
RUO
Primary
6773
Reconstitute at 1 mg/mL in deionized water.
Immunoprecipitation, KnockDown, Western Blot
Western Blot 0.5 ug/mL, Immunoprecipitation 3 ug/10^6 cells, Knockout Validated
P52630
interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human STAT2 in Western blots.
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit