Learn More
Transglutaminase 4/TGM4 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF5760
Catalog No. AF5760
Add to cart
This item is not returnable. View return policy
Description
Description
Transglutaminase 4/TGM4 Polyclonal antibody AF5760 Specifically detects Transglutaminase 4/TGM4 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications
Specifications
Transglutaminase 4/TGM4
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
AAC50516
EC 2.3.2.13, fibrinoligase, hTGP, Prostate transglutaminase, Prostate-specific transglutaminase, protein-glutamine gamma-glutamyltransferase 4, TG(P), TGase P, TGase-4, TGM4, TGP, TGPFLJ26776, transglutaminase 4 (prostate), Transglutaminase P, Transglutaminase-4
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Transglutaminase 4 in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Transglutaminase 2, rhTransglutaminase 3, rhTransglutaminase 7, and recombinant mouse Transglutaminase 2 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TGM4
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Transglutaminase 4 Met2-Lys684 Accession # AAC50516
Immunogen affinity purified
RUO
Primary
7047
Reconstitute at 0.2 mg/mL in sterile PBS.