Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TrkA Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems AF175
This item is not returnable.
View return policy
Description
TrkA Polyclonal specifically detects TrkA in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF-ready.Specifications
TrkA | |
Polyclonal | |
Unconjugated | |
AAA36770 | |
NTRK1 | |
Mouse myeloma cell line NS0-derived recombinant human TrkA Ala33-Glu407 Accession # AAA36770 | |
100 μg | |
Primary | |
Detects human TrkA in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant rat TrkA is observed and 10% cross-reactivity with recombinant human (rh) TrkB and rhTrKC is observed. | |
Human | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Neutralization, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, Neutralization 3-12 ug/mL, CyTOF-ready | |
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, NTRK1, NTRK-1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
Goat | |
Affinity Purified | |
RUO | |
4914 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title