Learn More
Tyrosylprotein Sulfotransferase 2/TPST2 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF6236
Catalog No. AF6236
Add to cart
This item is not returnable. View return policy
Description
Description
Tyrosylprotein Sulfotransferase 2/TPST2 Polyclonal antibody AF6236 Specifically detects Tyrosylprotein Sulfotransferase 2/TPST2 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Tyrosylprotein Sulfotransferase 2/TPST2
LYOPH
Western Blot 0.5 ug/mL
O60704
EC 2.8.2.20, TPST-2, tyrosylprotein phosphotransferase 2, Tyrosylprotein Sulfotransferase 2, tyrosylprotein sulfotransferase 2protein-tyrosine sulfotransferase 2, tyrosylprotein sulfotransferase-2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Tyrosylprotein Sulfotransferase 2/TPST2 in direct ELISAs and Western blots. In direct ELISAs, less than 7% cross-reactivity with recombinant human TPST1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TPST2
Chinese hamster ovary cell line CHO-derived recominant human Tyrosylprotein Sulfotransferase 2/TPST2 Gln26-Ser377 (predicted) Accession # O60704
Immunogen affinity purified
RUO
Primary
8459
Reconstitute at 0.2 mg/mL in sterile PBS.