Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human UbcH2/UBE2H, Polyclonal, R&D Systems™

Rabbit Polyclonal Antibody

Manufacturer:  R&D Systems A605

Catalog No. A605


Add to cart
This item is not returnable. View return policy

Description

Description

Host Species: Rabbit
Species Reactivity:Human

Applications:

Western Blotting

Specifications

Specifications

UbcH2/UBE2H
Unconjugated
Supplied in a solution of PBS and Glycerol. with No Preservative
E2-14K, E2-20K, EC 6.3.2.19, UBC8, UBC8E2-20K, UBCH, UBCH2, UBE2H, Ubiquitin carrier protein H, ubiquitin-conjugating enzyme E2 H, Ubiquitin-conjugating enzyme E2-20K, ubiquitin-conjugating enzyme E2H (homologous to yeast UBC8), ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast), Ubiquitin-protein ligase H
Rabbit
IgG
RUO
Primary
7328
Human
Western Blot
Western Blot
Affinity Purified
UBE2H
Full-length, highly purified human UbcH2 protein
200 uL
Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Polyclonal
Detects UbcH2/UBE2H in Western blots.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit