Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human Ubiquitin, Polyclonal, R&D Systems™

Rabbit Polyclonal Antibody

Manufacturer:  R&D Systems A100

Catalog No. A100


Add to cart
This item is not returnable. View return policy

Description

Description

Host Species: Rabbit
Species Reactivity:Human

Applications:

Western Blotting

Specifications

Specifications

Ubiquitin
Unconjugated
Supplied as a solution in PBS. with No Preservative
RPS27A, UBA52, UBB, UBB ubiquitin B, UBC
Rabbit
IgG
200 uL
Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Polyclonal
The antibody cross-reacts with mouse, rat and human ubiquitin; but not with Nedd8 or Sumo1 proteins. This reagent is non-functional for immunoprecipitation.
Western Blot
Western Blot
Affinity Purified
UBB
Purified human recombinant ubiquitin
Antigen Affinity-purified
RUO
Primary
6233
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit