Learn More
Pepro Tech Inc Anti-Human Uteroglobin, 100ug DFS Item
Manufacturer:  Pepro Tech Inc 500P330100UG
Produced from sera of rabbits immunized with highly pure Recombinant Human Uteroglobin. Anti-Human Uteroglobin-specific antibody was purified by affinity chromatography employing an immobilized Human Uteroglobin matrix.
Catalog No. 50-195-2603