Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Human XCR1, PE, Polyclonal, R&D Systems™

Manufacturer:  R&D Systems FAB857P025

 View more versions of this product

Catalog No. FAB857P025


Add to cart

Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Hu
Immunogen: E. coli-derived recombinant human XCR1. Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267

Applications:

Flow Cytometry

Specifications

Specifications

XCR1/CCXCR1
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Affinity Purified
XCR1
E. coli-derived recombinant human XCR1, Met1-Thr31, Ser89-Lys103, His168-His190, Phe251-Tyr267, Accession # P46094
Antigen Affinity-purified
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Polyclonal
Detects human XCR1 in direct ELISAs and Western blots.
Flow Cytometry
Flow Cytometry 10 uL/10^6 cells
P46094
CCXCR1, CCXCR1XC chemokine receptor 1, chemokine (C motif) receptor 1, chemokine XC receptor 1, G protein-coupled receptor 5, GPR5, GPR5chemokine (C motif) XC receptor 1, G-protein coupled receptor 5, Lymphotactin receptor
Goat
IgG
25 Tests
Primary
2829
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit