Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Humanin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA146256
Description
Humanin Polyclonal antibody specifically detects Humanin in Human samples. It is validated for Western Blot.Specifications
Humanin | |
Polyclonal | |
Unconjugated | |
MT-RNR2 | |
BCL2L4; HN1; Humanin-like 1; humanin-like protein 1; mitochondrially encoded 16S RNA; MTRNR2; MT-RNR2; MTRNR2L1; MT-RNR2-like 1; MTRNR2-like 1; MT-RNR2-like protein 1; RNR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4550 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8IVG9 | |
MT-RNR2 | |
Residues 9-24 of the human Humanin protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction