Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
hormonally up-regulated Neu-associated kinase, Mouse, Clone: 2G3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant HUNK.
Supplier: Abnova Corporation H00030811M03
Description
Sequence: LHPTLVSFAHEDKNSPPKEEGLCCPPPVPSNGPMQPLGSPNCVKSRGRFPMMGIGQMLRKRHQSLQPSADRPLEASLPPLQPLAPVNLAFDMADGVKTQCSpecifications
hormonally up-regulated Neu-associated kinase | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_014586 | |
Mouse | |
Affinity Purified | |
RUO | |
Yes | |
30811 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |
ELISA, Western Blot | |
2G3 | |
Mouse monoclonal antibody raised against a partial recombinant HUNK. | |
HUNK | |
HUNK | |
HUNK (NP_055401, 615 a.a. ∼ 714 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Kinases | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction