Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Huntingtin Mouse anti-Human, Mouse, Clone: 1A771, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA141172

Catalog No. 01-676-481

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Huntington's disease (HD) is a neurodegenerative disorder caused by an expanding polyglutamine repeat in the huntingtin gene. HD is a mid-life onset autosomal dominant neurodegenerative disease that is characterized by psychiatric disorders, dementia, and involuntary movements (chorea), leading to death in 10-20 years. The HD gene product is widely expressed in human tissues, with the highest level of expression in the brain. The huntington gene product is expressed at similar levels in patients and controls, which suggests that the expansion of the polyglutamine repeat induces a toxic gain of function perhaps through interactions with other cellular proteins. Using yeast two-hybrid system Li et al., have identified a protein, HAP1 (huntington associated protein 1), that associates with huntingtin protein. The in vitro data suggest that the association between HAP1 and huntington is enhanced by increasing length of glutamine repeat.

Clone 1A771 binds to the expanded polyQ repeat form of Htt in WB, showing no detectable binding to normal Htt in WB.Suggested positive control: Lymphoblast cell line extracts from HD patients.
Specifications

Specifications

Huntingtin
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P42858, P42859
HD, IT15
Mouse
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
15194, 3064
Western Blot
1A771
Unconjugated
HTT
Liquid
HTT
Fusion protein containing a repeating sequence of 62 consecutive glutamine residues.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit