Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-HUWE1, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP145233

Catalog No. NBP145233


Add to cart

Description

Description

HUWE1 Polyclonal antibody specifically detects HUWE1 in Human, Mouse samples. It is validated for Peptide ELISA.
Specifications

Specifications

HUWE1
0.5 mg/mL
Peptide ELISA Detection limit 1:128000
NP_113584.3
EC 6.3.2, EC 6.3.2.6, HECT, UBA and WWE domain containing 1, HECTH9, HSPC272, KIAA0312MULE, Mcl-1 ubiquitin ligase E3, Mule, UREB1UBA and WWE domain-containing protein 1, URE-binding protein 1
Goat
IgG
0.1 mg
Stem Cell Markers
Primary
10075
Human, Mouse
ELISA
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
HUWE1
Peptide with sequence C-DDLKSNTEYHKYQ corresponding to internal region according to NP_113584.3.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody recognizes and epitope within the internal region.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only