Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYAL1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527901

Catalog No. PIPA527901


Add to cart

Description

Description

PA5-27901 targets HYAL1 in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-27901 immunogen is recombit fragment corresponding to a region within amino acids 3 and 374 of Human HYAL1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes a lysosomal hyaluronidase. Hyaluronidases intracellularly degrade hyaluro, one of the major glycosaminoglycans of the extracellular matrix. Hyaluro is thought to be involved in cell proliferation, migration and differentiation. This enzyme is active at an acidic pH and is the major hyaluronidase in plasma. Mutations in this gene are associated with mucopolysaccharidosis type IX, or hyaluronidase deficiency. The gene is one of several related genes in a region of chromosome 3p21.3 associated with tumor suppression. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

HYAL1
Polyclonal
Unconjugated
HYAL1
Liquid
HYAL1
Recombit fragment corresponding to a region within amino acids 3 and 374 of Human HYAL1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q12794
HYAL-1, LUCA1, NAT6, hyaluronidase 1, hyaluronidase-1, hyaluronoglucosaminidase-1, luCa-1, lung carcinoma protein 1, plasma hyaluronidase, tumor suppressor LUCA-1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3373
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit