Learn More
HYOU1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant HYOU1.
Supplier: Abnova Corporation H00010525A02
Description
The protein encoded by this gene belongs to the heat shock protein 70 family. This gene uses alternative transcription start sites. A cis-acting segment found in the 5' UTR is involved in stress-dependent induction, resulting in the accumulation of this protein in the endoplasmic reticulum (ER) under hypoxic conditions. The protein encoded by this gene is thought to play an important role in protein folding and secretion in the ER. Since suppression of the protein is associated with accelerated apoptosis, it is also suggested to have an important cytoprotective role in hypoxia-induced cellular perturbation. This protein has been shown to be up-regulated in tumors, especially in breast tumors, and thus it is associated with tumor invasiveness. This gene also has an alternative translation initiation site, resulting in a protein that lacks the N-terminal signal peptide. This signal peptide-lacking protein, which is only 3 amino acids shorter than the mature protein in the ER, is thought to have a housekeeping function in the cytosol. In rat, this protein localizes to both the ER by a carboxy-terminal peptide sequence and to mitochondria by an amino-terminal targeting signal. [provided by RefSeq]
Sequence: EVQYLLNKAKFTKPRPRPKDKNGTRAEPPLNASASDQGEKVIPPAGQTEDAEPISEPEKVETGSEPGDTEPLELGGPGAEPEQKEQSTGQKRPLKNDELSpecifications
HYOU1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant HYOU1. | |
HYOU1 | |
DKFZp686N08236/FLJ94899/FLJ97572/Grp170/HSP12A/ORP150 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Liquid |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_006389 | |
HYOU1 | |
HYOU1 (NP_006380, 901 a.a. ∼ 999 a.a) partial recombinant protein with GST tag. | |
RUO | |
10525 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.