Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IκBε Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611408

Catalog No. BDB611408

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NF-κB is a ubiquitously expressed transcription factor that regulates many cytokine and Ig genes. It consists of homo- and heterodimeric proteins that belong to the Rel family of transcription factors. Rel proteins such as p50, p52, p65 (RelA), c-Rel, and RelB contain a Rel homology region (RHR) that includes DNA-binding and dimerization domains and a nuclear localization signal. Inactive NF-κB complexes are associated with their inhibitory IκB proteins. Several IκB proteins have been characterized including IκBα, IκBβ, and IκBε. They contain ankyrin repeats that interact with Rel RHR regions to form complexes in the cytoplasm that prevent nuclear translocation of NF-κB. In response to stimuli such as cytokines, LPS, and viral infections, IκB proteins are phosphorylated at critical residues by IκB kinases (IKKα and IKKβ) leading to proteolysis of IκB proteins. This frees the heterodimeric NF-κB to form a heterotetramer that translocates to the nucleus and acts as a transcriptional activator. IκBε may function primarily in the cytoplasm where it sequesters p65 and c-Rel, while IκBα and IκBβ have additional functions that involve entering the nucleus to inhibit Rel DNA binding.

Host Species: Mouse
Clone: 21
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Human IκBε aa. 200-211
Formula Weight [Chemical]: 45kDa

Immunofluorescence, Western Blotting

Specifications

Specifications

IκBε
21
Unconjugated
Murine
IgG2a
RUO
Primary
Human, Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Human IκBε aa. 200-211
50μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit