Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IκBα Mouse anti-Human, Unlabeled, Clone: 25, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610690

 View more versions of this product

Catalog No. BDB610690


Add to cart

Description

Description

NF-κB is a transcription factor that regulates many cytokine and Ig genes involved in immune response, inflammation, and viral infections. NF-κB is induced by cytokines and lipopolysaccharide (LPS), but is inhibited by glucocorticoids. IκBα/MAD-3 inhibits the DNA-binding activity of NF-κB and related transcription factors. In THP-1 monocytic-like cells, LPS induces the phosphorylation and inactivation of IκBα, which results in the activation of NF-κB. Therefore, phosphorylation of IκBα may induce the dissociation of the IκBα/NF-κB complex.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

IκBα
25
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
MAD-3; I-kappa-B-alpha; IkB-alpha; NFKBIA; NF-kappa-B inhibitor alpha
Affinity Purified
Human IκBα aa. 145-302
50μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.