Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF2BP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524026

Catalog No. PIPA524026

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

IRF2BP2 encodes an interferon regulatory factor-2 (IRF2)binding protein that interacts with the C-terminal transcriptionalrepression domain of IRF2. Alternative splicing results in multipletranscript variants encoding distinct isoforms.
Specifications

Specifications

IRF2BP2
Polyclonal
PBS with 0.09% sodium azide
Q7Z5L9
IRF-2-binding protein 2, interferon regulatory factor 2-binding protein 2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
359948
Flow Cytometry, Western Blot
Unconjugated
IRF2BP2
Liquid
IRF2BP2
KLH conjugated synthetic peptide between 337-366 amino acids from the central region of human I2BP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit