Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IBTK, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP178061

Catalog No. NBP178061


Add to cart

Description

Description

IBTK Polyclonal antibody specifically detects IBTK in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

IBTK
1.63 mg/ml
Western Blot 1:500-1:3000, ELISA 1:5000-1:25000
Q9P2D0
Bruton agammaglobulinemia tyrosine kinase inhibitor, BTK-binding protein, BTKI, DKFZP564B116, IBtk, inhibitor of Bruton agammaglobulinemia tyrosine kinase, inhibitor of Bruton tyrosine kinase, inhibitor of Bruton's tyrosine kinase, inhibitor of Bruton's tyrosine kinase-alpha, inhibitor of Bruton's tyrosine kinase-beta, inhibitor of Bruton's tyrosine kinase-gamma, KIAA1417, MGC142256, MGC142258, RP1-93K22.1
Rabbit
IgG
0.1 mg
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
ELISA, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
IBTK
This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to aa 1205-1217 of Human IBTK. IBTK is a currently uncharacterized human ankyrin repeat protein BAA92655. This protein has been suggested to bind to CUL3.
Immunogen affinity purified
RUO
Primary
25998
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only