Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBTK Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524224

Catalog No. PIPA524224

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene binds to Bruton ft.s tyrosine kinase (BTK) and downregulates BTK ft.s kinase activity. In addition, the encoded protein disrupts BTK-mediated calcium mobilization and negatively regulates the activation of nuclear factor-kappa-B-driven transcription.
Specifications

Specifications

IBTK
Polyclonal
PBS with 0.09% sodium azide
Q9P2D0
BTKI, RP1-93K22.1, BTK-binding protein, Bruton agammaglobulinemia tyrosine kinase inhibitor, inhibitor of Bruton tyrosine kinase, inhibitor of Bruton′s tyrosine kinase, inhibitor of Bruton′s tyrosine kinase-alpha, inhibitor of Bruton′s tyrosine kinase-beta, inhibitor of Bruton′s
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25998
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
IBTK
Liquid
IBTK
KLH conjugated synthetic peptide between 586-613 amino acids from the central region of human IBTK
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit