Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICBP90/UHRF1 Mouse anti-Human, Clone: 1RC-1C10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE308
Description
Anti-ICBP90/UHRF1 Antibody, clone 1RC1C-10 is a Mouse Monoclonal Antibody for detection of ICBP90/UHRF1 also known as E3 ubiquitin-protein ligase, inverted CCAAT box-binding protein and has been validated in western blot, Chimmunoprecipitation, immunocytochemistry and immunohistochemistry.
Specifications
ICBP90/UHRF1 | |
Monoclonal | |
Unconjugated | |
NP_001041666 | |
Mouse | |
Unpurified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |
ChIP Assay, Immunocytochemistry, Immunohistochemistry, Western Blot | |
1RC-1C10 | |
Mouse monoclonal IgG1 ascites with 0.05% sodium azide. | |
UHRF1, ICBP90, NP95, RNF106 | |
Recombinant protein corresponding to human ICBP90. | |
100 μL | |
Human | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction