Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICEBERG, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA128392

Catalog No. PIPA128392

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-28392 detects ICEBERG from human, rat samples.PA1-28392 has been successfully used in immunoprecipitation, Western blot applications.The PA1-28392 immunogen is: Synthetic peptide (Human).
Specifications

Specifications

ICEBERG
Polyclonal
Unconjugated
CARD18
Purified
CARD18
Synthetic peptide (Human).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Rat
ELISA, Immunoprecipitation, Western Blot
0.5mg/mL
PBS with 0.02% sodium azide
P57730
ICEBERG, UNQ5804, pseudo-ICE
Rabbit
IgG
50μg
Store at -20°C
Primary
59082
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit