Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IPCEF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524261

Catalog No. PIPA524261


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

Enhances the promotion of guanine-nucleotide exchange by PSCD2 on ARF6 in a concentration-dependent manner.
Specifications

Specifications

IPCEF1
Polyclonal
PBS with 0.09% sodium azide
Q8WWN9
PIP3-E, RP3-402L9.2, interaction protein for cytohesin exchange factors 1 Interaction protein for cytohesin exchange factors 1, interactor protein for cytohesin exchange factors 1, phosphoinositide binding protein PIP3-E, phosphoinositide-binding protein PIP3-E
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26034
Flow Cytometry, Western Blot
Unconjugated
IPCEF1
Liquid
IPCEF1
KLH conjugated synthetic peptide between 313-341 amino acids from the C-terminal region of human ICEF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit