Learn More
ICK Rabbit anti-Bovine, Human, Non-human primate, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA523161
Catalog No. PIPA523161
Add to cart
Description
Description
ICK is a 632 amino acid protein, belonging to the CMGC Ser/Thr protein kinase family, containing a single protein kinase domain. It is an intestinal serine/threonine kinase harboring a dual phosphorylation site found in mitogen-activating protein (MAP) kinases. It localizes to the intestinal crypt region and plays important role in intestinal epithelial cell proliferation and differentiation and is also needed for cardiac development. ICK is expressed in heart, brain, placenta, pancreas, thymus, prostate, testis, ovary, small intestine, colon and in many cancer cell lines.
Specifications
Specifications
ICK
Polyclonal
whole serum with 50% glycerol and 0.025% sodium azide
Q9UPZ9
ECO, LCK2, MRK, MAK-related kinase, hICK, laryngeal cancer kinase 2, serine/threonine protein kinase, serine/threonine-protein kinase ICK
Rabbit
IgG
RUO
Antibody
Polyclonal
Bovine, Human, Non-human Primate
Western Blot
Unconjugated
ICK
Liquid
ICK
Synthetic peptide (390 LEHKNGEIKPKSRRR 404) of ICK protein.
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
22858, 506286