Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICOS Mouse anti-Human, APC, Clone: 669222, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523680

Catalog No. PIMA523680


Add to cart

Description

Description

CD278 belongs to the CD28 and CTLA-4 cell-surface receptor family and is highly expressed in activated T-cells. It is essential both for efficient interaction between T and B-cells and for normal antibody responses to T-cell dependent antigens. It enhances all basic T-cell responses to a foreign antigen, namely proliferation, secretion of lymphokines, up-regulation of molecules that mediate cell-cell interaction, and effective help for antibody secretion by B-cells.
Specifications

Specifications

ICOS
Monoclonal
APC
ICOS
Liquid
ICOS
NS0 mouse myeloma cell line transfected with human ICOS and mouse myeloma cell line NS0-derived recombit human ICOS cocktail Glu21-Phe141
Protein A/G
RUO
Antibody
Monoclonal
Human
Flow Cytometry
669222
Buffered saline solution with BSA and 0.09% sodium azide
Q9Y6W8
AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator
Mouse
IgG1
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
29851
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit