Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF8, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131665

Catalog No. PIPA131665

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Interferon consensus sequence-binding protein (ICSBP) is a transcription factor of the interferon (IFN) regulatory factor (IRF) family. Proteins of this family are composed of a conserved DNA-binding domain in the N-terminal region and a divergent C-term

PA1-31665 detects ICSBP from human samples.PA1-31665 has been successfully used in Western blot applications.The PA1-31665 immunogen is: Peptide with sequence QRSFFRENQQITV, from C Terminus of the protein sequence according to NP_002154
Specifications

Specifications

ICSBP
Polyclonal
Unconjugated
IRF8
Purified
IRF8
Peptide with sequence QRSFFRENQQITV, from C Terminus of the protein sequence according to NP_002154
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q02556, P23611
H-ICSBP, ICSBP, ICSBP1, IRF-8
Goat
IgG
50μg
Avoid Freeze/Thaw Cycles
Primary
3394
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit