Learn More
ID1 Mouse anti-Human, Clone: 3A9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519169
Catalog No. PIMA519169
Add to cart
Description
Description
Peptide Sequence: MKVASGSTAT AAAGPSCALK AGKTASGAGE VVRCLSEQSV AISRCAGGAG ARLPALLDEQ QVNVLLYDMN GCYSRLKELV PTLPQNRKVS KVEILQHVID YIRDLQLELN SESEVGTPGG RGLPVRAPLS TLNGEISALT AEAACVPADD RILCR
The protein encoded by this gene is a helix-loop-helix (HLH) protein that can form heterodimers with members of the basic HLH family of transcription factors. The encoded protein has no DNA binding activity and therefore can inhibit the DNA binding and transcriptional activation ability of basic HLH proteins with which it interacts. This protein may play a role in cell growth, senescence, and differentiation. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
ID1
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
P41134
BHLHB24, bHLHb24DNA-binding protein inhibitor ID-1, Class B basic helix-loop-helix protein 24, dJ857M17.1.2, dJ857M17.1.2 (inhibitor of DNA binding 1, domit negative helix-loop-helixprotein), ID, inhibitor of differentiation 1, Inhibitor of DNA binding 1, inhibitor of DNA binding 1, domit negative helix-loop-helix protein
Mouse
IgG2a, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3397
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
3A9
Unconjugated
ID1
Liquid
ID1
ID1 (AAH00613.1, 1 a.a. ∽ 155 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human