Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

inhibitor of DNA binding 3, dominant negative helix-loop-helix protein, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human ID3 protein.

Manufacturer:  Abnova Corporation H00003399B01

Catalog No. 89-005-586


Add to cart

Description

Description

Members of the ID family of helix-loop-helix (HLH) proteins lack a basic DNA-binding domain and inhibit transcription through formation of nonfunctional dimers that are incapable of binding to DNA.[supplied by OMIM

Sequence: MKALSPVRGCYEAVCCLSERSLAIARGRGKGPAAEEPLSLLDDMNHCYSRLRELVPGVPRGTQLSQVEILQRVIDYILDLQVVLAEPAPGPPDGPHLPIQTAELAPELVISNDKRSFCH
Specifications

Specifications

inhibitor of DNA binding 3, dominant negative helix-loop-helix protein
Unconjugated
No additive
BC003107
HEIR-1|bHLHb25
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human ID3 protein.
ID3
Serum
ID3
ID3 (AAH03107, 1 a.a. ~ 119 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3399
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit