Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDH3G Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526798

Catalog No. PIPA526798

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predomitly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the gamma subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. This gene is a candidate gene for periventricular heterotopia.
Specifications

Specifications

IDH3G
Polyclonal
PBS with 0.09% sodium azide
P51553
H-IDHG, IDH-gamma, NAD (H)-specific isocitrate dehydrogenase gamma subunit, NAD(+)-specific ICDH subunit gamma, NAD+-specific ICDH, isocitrate dehydrogenase [NAD] subunit gamma, mitochondrial, isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit, iso
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3421
Flow Cytometry, Western Blot
Unconjugated
IDH3G
Liquid
IDH3G
KLH conjugated synthetic peptide between 366-393 amino acids from the C-terminal region of human IDH3G
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit