Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interferon, gamma-inducible protein 16, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human IFI16 protein.

Manufacturer:  Abnova Corporation H00003428B01

Catalog No. 89-005-592


Add to cart

Description

Description

This gene encodes a member of the HIN-200 (hematopoietic interferon-inducible nuclear antigens with 200 amino acid repeats) family of cytokines. The encoded protein contains domains involved in DNA binding, transcriptional regulation, and protein-protein interactions. The protein localizes to the nucleoplasm and nucleoli, and interacts with p53 and retinoblastoma-1. It modulates p53 function, and inhibits cell growth in the Ras/Raf signaling pathway. [provided by RefSeq

Sequence: MGKKYKNIVLLKGLEVINDYHFRMVKSLLSNDLKLNLKMREEYDKIQIADLMEEKFRGDAGLGKLIKIFEDIPTLEDLAETLKKEKLKVKGPALSRKRKKEVDATSPAPSTSSTVKTEGAEATPGAQKRKKSTKEKAGPKGSKVSEEQTQPPSPAGAGMSTAMGRSPSPKTSLSAPPNTSSTENPKTVAKCQVTPRRNVLQKRPVIVKVLSTTKPFEYETPEMEKKIMFHATVATQTQFFHVKVLNTSLKEKFNGKKIIIISDYLEYDSLLEVNEESTVSEAGPNQTFEVPNKIINRAKETLKIDILHKQASGNIVYGVFMLHKKTVNQKTTIYEIQDDRGKMDVVGTGQCHNIPCEEGDKLQLFCFRLRKKNQMSKLISEMHSFIQIKKKTNPRNNDPKSMKLPQEQSQLPNPSEASTTFPESHLRTPQMPPTTPSSSFFTKKSEDTISKMNDFMRMQILKEGSHFPGPFMTSIGPAESHPHTPQMPPSTPSSSFLTTLKPRLKTEPEEVSIEDSAQSDLKEVMVLNATESFVYEPKEQKKMFHATVATENEVFRVKVFNIDLKEKFTPKKIIAIANYVCRNGFLEVYPFTLVADVNADRNMEIPKGLIRSASVTPKINQLCSQTKGSFVNGVFEVHKKNVRGEFTYYEIQDNTGKMEVVVHGRLTTINCEEGDKLKLTCFELAPKSGNTGELRSVIHSHIKVIKTRKNKKDILNPDSSMETSPDFFF
Specifications

Specifications

interferon, gamma-inducible protein 16
Unconjugated
No additive
BC017059.1
IFNGIP1|MGC9466|PYHIN2
Mouse
50 uL
Transcription Regulation
Y
Polyclonal
Human
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human IFI16 protein.
IFI16
Serum
IFI16
IFI16 (AAH17059.1, 1 a.a. ~ 729 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3428
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit