Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IFIT3, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP131722

Catalog No. NBP131722


Add to cart

Description

Description

IFIT3 Polyclonal antibody specifically detects IFIT3 in Human samples. It is validated for Western Blot.
Specifications

Specifications

IFIT3
1.0mg/mL
Western Blot 1:500-1:3000
O14879
CIG-49, GARG-49, IFI60Retinoic acid-induced gene G protein, IFIT-3, IFIT-4, IFIT4IFI-60K, interferon-induced protein with tetratricopeptide repeats 3, Interferon-induced protein with tetratricopeptide repeats 4Interferon-induced 60 kDa protein, IRG2, ISG60, ISG-60, RIG-GCIG49
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
3437
Human
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Affinity Purified
IFIT3
Recombinant protein encompassing a sequence within the center region of human IFIT3. The exact sequence is proprietary.
56 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only