Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFITM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520988
Description
A suggested positive control is Jurkat cell lysate. PA5-20988 can be used with blocking peptide PEP-1102.
IFITM1 Polyclonal antibody specifically detects IFITM1 in Human samples. It is validated for Western Blot.Specifications
IFITM1 | |
Polyclonal | |
Unconjugated | |
Ifitm1 | |
1110036C17Rik; 27-Sep; 9-27; CD225; dispanin subfamily A member 2a; DSPA2a; Fragilis protein 2; fragilis2; IFI17; IFITM1; ifitm-like protein 2; interferon induced transmembrane protein 1; interferon induced transmembrane protein 2 like; interferon-induced protein 17; Interferon-induced transmembrane protein 1; interferon-inducible protein 9-27; LEU13; Leu-13 antigen; Mil2; Mil-2; Mouse ifitm-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8519 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P13164 | |
Ifitm1 | |
A 14 amino acid peptide near the carboxy terminus of human IFITM1. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction