Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFITM3/Fragilis Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | IFITM3/Fragilis |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5-2 ug/ml, Simple Western 1:10 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB6001534SS
|
Novus Biologicals
NB6001534SS |
0.025 mg |
Each for $204.00
|
|
|||||
NB6001534
|
Novus Biologicals
NB6001534 |
0.1 mg |
Each for $409.50
|
|
|||||
Description
IFITM3/Fragilis Polyclonal specifically detects IFITM3/Fragilis in Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
IFITM3/Fragilis | |
Western Blot 0.5-2 ug/ml, Simple Western 1:10 | |
Polyclonal | |
Purified | |
RUO | |
PBS containing 0.05% BSA with 0.05% Sodium Azide | |
1-8U, interferon induced transmembrane protein 3 (1-8U), interferon-induced transmembrane protein 3, Interferon-inducible protein 1-8U, IP15 | |
IFITM3 | |
IgG | |
Protein G purified | |
Fragilis - Germ Line Marker |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat | |
Q9CQW9 | |
10410 | |
A portion of amino acids 1-50 of mouse fragilis was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title