Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFITM3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511275

Catalog No. PIPA511275

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The family of interferon-induced transmembrane protein (Ifitm/mil/fragilis) cell surface proteins may modulate cell adhesion and influence cell differentiation.
Specifications

Specifications

IFITM3
Polyclonal
PBS with 0.09% sodium azide
Q01628
Interferon-induced transmembrane protein 3; Interferon-inducible protein 1-8U
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10410
Western Blot
Unconjugated
IFITM3
Liquid
IFITM3
KLH conjugated synthetic peptide between 64-93 amino acids from the central region of human Fagilis (IFITM3)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit