Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IFN-alpha, Clone: K9, R&D Systems™
Mouse Monoclonal Antibody has been used in 8 publications
Supplier: R&D Systems 271001
This item is not returnable.
View return policy
Description
IFN-alpha Monoclonal specifically detects IFN-alpha in Porcine samples. It is validated for Neutralization.Specifications
IFN-alpha | |
Monoclonal | |
Unconjugated | |
Supplied as a liquid with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3440 | |
Porcine | |
Purified |
Neutralization | |
K9 | |
Neutralization | |
IFNA, IFNA2, IFNA2b, IFNalpha, IFN-alphaA, INFA2, interferon alpha 2, LeIF D | |
Porcine IFN-alpha | |
500 μg | |
Primary | |
Neutralizes pig interferon alpha | |
After receipt, this product should be kept at -20degreesC for retention of full activity. Do not store below -20degreesC. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title