Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN gamma Mouse anti-Rat, Clone: 88928, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523762

Catalog No. PIMA523762


Add to cart

Description

Description

In sandwich ELISAs, observed cross-reactivities were less than 2% with recombit mouse IFN- gamma and less than 0.1% with recombit human IFN- gamma and recombit porcine IFN- gamma. Reconstitute at 0.5 mg/mL in sterile PBS.

IFN gamma (Interferon gamma, Type II interferon) is a macrophage activation factor, and immune interferon that is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a dimeric protein consisting of two 146 amino acid subunits. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE, IFN gamma appears as a combination of 25, 20 and minor 15.5 kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific. Human IFN gamma does not show cross-reactivity with mouse. IFN gamma function includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFN gamma is involved in cytokine regulation and also acts synergistically with other cytokines. Activation of IFN gamma takes place through binding of IFN gamma receptor I and II, and activating the JAK-STAT pathway. IFN gamma does not show any homology with IFN alpha or IFN beta but human IFN gamma shows about 40% sequence homology with mouse IFN gamma. IFN gamma is upregulated by IL2, FGF basic, EGF and downregulated by vitamin D3 or DMN. IFN gamma gene mutations are associated with aplastic anemia.
Specifications

Specifications

IFN gamma
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P01581
IFN-gamma, IFN-y, Interferon y, IFNG
Mouse
IgG1
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
25712
ELISA, Immunocytochemistry, Immunofluorescence
88928
Unconjugated
Ifng
Lyophilized
IFNG
E. coli-derived recombit rat IFN- gamma Gln23-Cys156
Protein A/G
RUO
Antibody
Monoclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit