Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-IFN gamma, FITC, Clone: XMG1.2,
Rat Monoclonal Antibody
$269.00
Specifications
Antigen | IFN gamma |
---|---|
Clone | XMG1.2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | FITC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1518MI
|
MilliporeSigma™
MABF1518 |
100 μg |
Each of 1 for $269.00
|
|
|||||
Description
Specifically detects IFN gamma Clone: XMG1.2 in Mouse samples, and it is validated for Flow Cytometry
The XMG1.2 antibody is specific for mouse Interferon-gamma (IFN-g), a 20kDa type II cytokine known for its central roles in protection against bacterial or viral pathogens and for its anti-tumor properties. IFN-g is secreted by several types of immune cells, which allow the cytokine to modulate innate immunity, when secreted by NK and NKT cells, and to function in support of adaptive immunity when secreted by Th1 and CD8+ T cells (CTLs).The XMG1.2 antibody is suitable for detection of intracellular IFN-g protein, e.g. by flow cytometry, as well as for quantitative analysis of the secreted protein by ELISA, when paired with an appropriate secondary antibody. This clone is also widely used for neutralization of the functional activity of IFN-g in a variety of assays.Specifications
IFN gamma | |
Flow Cytometry | |
FITC | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
Ifng | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
XMG1.2 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P01580 | |
Proprietary immunogen | |
Purified by size-exclusion chromatography |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title