Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interferon, alpha 17, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human IFNA17 protein.

Manufacturer:  Abnova Corporation H00003451B01

Catalog No. 89-005-599


Add to cart

Description

Description

Sequence: MALSFSLLMAVLVLSYKSICSLGCDLPQTHSLGNRRALILLAQMGRISPFSCLKDRHDFGLPQEEFDGNQFQKTQAISVLHEMIQQTFNLFSTEDSSAAWEQSLLEKFSTELYQQLNNLEACVIQEVGMEETPLMNEDSILAVRKYFQRITLYLTEKKYSPCAWEVVRAEIMRSLSFSTNLQKILRRKD
Specifications

Specifications

interferon, alpha 17
Unconjugated
No additive
NM_021268
IFNA|INFA|LEIF2C1
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
3451
Western Blot
Mouse polyclonal antibody raised against a full-length human IFNA17 protein.
IFNA17
Serum
IFNA17
IFNA17 (NP_067091, 1 a.a. ~ 189 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit