Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFRD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | IFRD1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1.0-4.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224679SS
|
Novus Biologicals
NBP224679SS |
0.025 mg |
Each for $204.00
|
|
|||||
NBP224679
|
Novus Biologicals
NBP224679 |
0.1 mg |
Each for $409.50
|
|
|||||
Description
IFRD1 Polyclonal specifically detects IFRD1 in Human, Porcine, Bovine, Canine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IFRD1 | |
Western Blot 1.0-4.0 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
3475 | |
A portion of amino acids 70-120 of human Tis7 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Porcine, Bovine, Canine, Primate | |
12-O-tetradecanoylphorbol-13-acetate-induced sequence 7, interferon-related developmental regulator 1, Nerve growth factor-inducible protein PC4, PC4, pheochromocytoma cell-4, TIS7, TPA induced sequence 7 | |
IFRD1 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human, cow, horse, dog, chimpanzee, 92% homologous in mouse, rat, macaque, rhesus monkey, and 83% homologous in opossum. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title