Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFT81 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524017

Catalog No. PIPA524017

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency.Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis.
Specifications

Specifications

IFT81
Polyclonal
PBS with 0.09% sodium azide
Q8WYA0, O35594
CDV-1, CDV-1R, CDV1, CDV1R, carnitine deficiency-associated gene expressed in ventricle 1, carnitine deficiency-associated protein expressed in ventricle 1, intraflagellar transport protein 81 homolog
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12589, 28981
Western Blot
Unconjugated
IFT81
Liquid
IFT81
KLH conjugated synthetic peptide between 527-555 amino acids from the C-terminal region of human IFT81
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit